Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106343 Supplier Invitrogen™ Supplier No. PA5106343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BBS7.

BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BBS7
Gene Accession No. Q8IWZ6, Q8K2G4
Gene Alias 8430406N16Rik; Bardet-Biedl syndrome 7; Bardet-Biedl syndrome 7 (human); Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; Bbs2l1; BBS2-like 1; BBS2-like protein 1; BBS7; FLJ10715
Gene Symbols BBS7
Host Species Rabbit
Immunogen A synthesized peptide derived from human BBS7(Accession Q8IWZ6), corresponding to amino acid residues E399-A449.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361930, 55212, 71492
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.