Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116767 Supplier Invitrogen™ Supplier No. PA5116767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat testis, Rat lung Immunogen sequence: MDLILNRMDY LQVGVTSQKT MKLIPASRHR ATQKVVIGDH DGVVMCFGMK KGEAAAVFKT LPGPKIARLE LGGVINTPQE KIFIAAASEI RGFTKRGKQF LSFETNLTES IKAMHISGSD LFLSASYIYN HYCDCKDQHY YLSGDKINDV ICLPVERLSR ITPVLACQDR VLRVLQGSDV MYAVEVPGPP TVLALHNGNG GDSGEDLLFG TSDGKLALIQ ITTSKPVRKW EIQNEKKRGG ILCIDSFDIV GDGVKDLLVG RDDGMVEVYS.

BBS7, also named as BBS2L1, is a component of the BBSome complex which is required for ciliogenesis but is dispensable for centriolar satellite function. BBS7 is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BBS7
Gene Accession No. Q8IWZ6, Q8K2G4
Gene Alias 8430406N16Rik; Bardet-Biedl syndrome 7; Bardet-Biedl syndrome 7 (human); Bardet-Biedl syndrome 7 protein; Bardet-Biedl syndrome 7 protein homolog; Bbs2l1; BBS2-like 1; BBS2-like protein 1; BBS7; FLJ10715
Gene Symbols BBS7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human BBS7 (NP_0606602).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361930, 55212, 71492
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.