Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554373 Supplier Invitrogen™ Supplier No. PA554373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEDTQELGWE ETVDAAISHL LKTCLSKSSK EQALNLNSQL NIPKDTSQLK KHITLLCDRL SKGGRLCLST DAAAPQTMVM PGGCTTIPES DLEERSVEQD STELFTNHRH LTAETPRPEV SPLQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. Required for proper BBSome complex assembly and its ciliary localization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BBS9
Gene Accession No. Q3SYG4
Gene Alias B1; Bardet-Biedl syndrome 9; Bardet-Biedl syndrome 9 (human); bardet-Biedl syndrome 9 protein; Bardet-Biedl syndrome 9 protein homolog; BBS9; C18; D1; E130103I17Rik; EST 3159894; parathyroid hormone-responsive B1 gene protein; Parathyroid hormone-responsive B1 gene protein homolog; protein PTHB1; PTHB1; PTH-responsive osteosarcoma B1 protein; RGD1308472
Gene Symbols BBS9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BBS9. Recombinant protein control fragment (Product #RP-92689).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.