Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BBS9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100220
Description
Antibody detects endogenous levels of total BBS9.
The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. Required for proper BBSome complex assembly and its ciliary localization.
Specifications
BBS9 | |
Polyclonal | |
Unconjugated | |
BBS9 | |
B1; Bardet-Biedl syndrome 9; Bardet-Biedl syndrome 9 (human); bardet-Biedl syndrome 9 protein; Bardet-Biedl syndrome 9 protein homolog; BBS9; C18; D1; E130103I17Rik; EST 3159894; parathyroid hormone-responsive B1 gene protein; Parathyroid hormone-responsive B1 gene protein homolog; protein PTHB1; PTHB1; PTH-responsive osteosarcoma B1 protein; RGD1308472 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27241, 319845 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q3SYG4, Q811G0 | |
BBS9 | |
A synthesized peptide derived from human BBS9(Accession Q3SYG4), corresponding to amino acid residues G767-D817. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction