Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS9 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551315

Catalog No. PA551315


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total PTHB1 / BBS9 protein.

The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. Required for proper BBSome complex assembly and its ciliary localization.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BBS9
Polyclonal
Unconjugated
BBS9
B1; Bardet-Biedl syndrome 9; Bardet-Biedl syndrome 9 (human); bardet-Biedl syndrome 9 protein; Bardet-Biedl syndrome 9 protein homolog; BBS9; C18; D1; E130103I17Rik; EST 3159894; parathyroid hormone-responsive B1 gene protein; Parathyroid hormone-responsive B1 gene protein homolog; protein PTHB1; PTHB1; PTH-responsive osteosarcoma B1 protein; RGD1308472
Rabbit
Antigen Affinity Chromatography
RUO
27241
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q3SYG4
BBS9
Fusion protein of human BBS9.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.