Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BBX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568584
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 100%.
BBX gene ontology annotations related to this gene include DNA binding.
Specifications
BBX | |
Polyclonal | |
Unconjugated | |
BBX | |
5530401J07Rik; 5730403O13Rik; Ag recognized by Treg cells 1; ARTC1; BBX; BBX high mobility group box domain containing; BBX, HMG-box containing; bobby sox homolog; bobby sox homolog (Drosophila); HBP2; HMG box transcription factor BBX; HMG box-containing protein 2; HMG-BOX transcription factor BBX; HSPC339; MDS001; x 001 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
56987 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WY36 | |
BBX | |
synthetic peptide directed towards the N-terminal region of human BBX. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction