Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552246 Supplier Invitrogen™ Supplier No. PA552246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSLPQYSPVT FDRKCVPVPR KKKKTGNVSS EPTKTSKGSG DKWSNKQLFL DAIHPTEAIF SEDRNTMEPV HKVKNIPSIF NTPEPTTTQE PLVGSQKRKA RKTKI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

BBX gene ontology annotations related to this gene include DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBX
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BBX
Gene Accession No. Q8WY36
Gene Alias 5530401J07Rik; 5730403O13Rik; Ag recognized by Treg cells 1; ARTC1; BBX; BBX high mobility group box domain containing; BBX, HMG-box containing; bobby sox homolog; bobby sox homolog (Drosophila); HBP2; HMG box transcription factor BBX; HMG box-containing protein 2; HMG-BOX transcription factor BBX; HSPC339; MDS001; x 001 protein
Gene Symbols BBX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BBX. Recombinant protein control fragment (Product #RP-102501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56987
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.