Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BC-LI-0186
GREENER_CHOICE

Catalog No. 679125
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-912-5 25 mg
67-915 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-912-5 Supplier Tocris Bioscience™ Supplier No. 6791/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Leucyl-tRNA synthase (TRS)/Ras-related GTP-binding protein D (RagD) interaction inhibitor

BC-LI-0186 is a potent and high affinity leucyl-tRNA synthase (TRS)/Ras-related GTP-binding protein D (RagD) interaction inhibitor (IC50 = 46.1 nM; Kd = 42.1 nM for binding to LRS). Inhibits mTOR complex 1 activity by blocking binding to LRS without affecting catalytic activity. Inhibits leucine-induced translocation of LRS to the lysosome.

Specifications

Chemical Name or Material 4-[2,5-Dihydro-2,3-dimethyl-4-(1-methylethyl)-5-oxo-1 H-pyrazol-1-yl]-N-(2-phenoxyethyl)benzenesulfonamide
CAS 695207-56-8
Quantity 25 mg
Target Ras GTPase Inhibitors
Molecular Formula C22H27N3O4S
Formula Weight 3939
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.