Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCA2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987475
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7475 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7475 Supplier Bioss Supplier No. BS6757R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF364 contains 1 RING-type zinc finger. The exact function of ZNF364 remains unknown.

Specifications

Antigen BCA2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RNF115
Gene Accession No. Q9Y4L5
Gene Alias 2610028E05Rik; AU042696; BCA2; BOS_3018; CL469780; E3 ubiquitin-protein ligase RNF115; LOC100357753; Rab7-interacting RING finger protein; Rabring 7; Rabring7; RING finger protein 115; RING-type E3 ubiquitin transferase RNF115; RNF115; Zfp364; Zinc finger protein 364; ZNF364
Gene Symbols RNF115
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BCA2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27246
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.