Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCA3 Antibody (OTI1E3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272057H
Description
BCA3 Monoclonal specifically detects BCA3 in Human samples. It is validated for Western Blot.Specifications
BCA3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
AKIP1 | |
Full length recombinant protein of human BCA3 (NP_878904) produced in E.coli. | |
0.1 mL | |
56672.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1E3 | |
Western Blot | |
AKIP1koyt binding protein 2, BCA3koyt binding protein 3, breast cancer associated gene 3, Breast cancer-associated gene 3 protein, chromosome 11 open reading frame 17, koyt binding protein 1, PKA-interacting protein, proline-rich protein BCA3, protein kinase A-interacting protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction