Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM/CD239 Antibody (002), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289356IR
Description
BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
BCAM/CD239 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
4059 | |
Human | |
Purified |
ELISA | |
2 | |
ELISA, Sandwich ELISA Detection | |
antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human BCAM/CD239 (NP_005572.2; Met1-Ala547). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human ALCAM Human CD31 Human EpCAM Human ICAM1 Human ICAM2 Human ICAM3 Human MCAM Human VCAM1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction