Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM/CD239 Antibody (MM0107-1M39), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212050JF646
Description
BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BCAM/CD239 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BCAM | |
Human recombinant BCAM/CD239 (extracellular domain) | |
0.1 mL | |
4059 | |
Store at 4°C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0107-1M39 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction