Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM/CD239 Antibody (SR2254), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | BCAM/CD239 |
---|---|
Clone | SR2254 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
BCAM/CD239 Monoclonal antibody specifically detects BCAM/CD239 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
BCAM/CD239 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
A synthesized peptide derived from human BCAM/CD239 (Uniprot #: P50895) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR2254 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, CD Markers, Immunology | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
4059 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title