Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM (CD239) Mouse anti-Human, Brilliant Violet 480, Clone: B64, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 748015
Description
BCAM (CD239) Monoclonal antibody specifically detects BCAM (CD239) in Human samples. It is validated for Flow Cytometry.
Specifications
BCAM (CD239) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
BCAM (CD239) | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
B64 | |
Brilliant Violet 480 | |
50 μg | |
Primary | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction