Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCAM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580374
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human BCAM.
Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
BCAM | |
Polyclonal | |
Unconjugated | |
BCAM | |
1200005K12Rik; antigen identified by monoclonal antibody F8; AU; Auberger B antigen; basal cell adhesion molecule; basal cell adhesion molecule (Lu and Au blood groups); basal cell adhesion molecule (Lutheran blood group); basal cell adhesion molecule Lutheran blood group; BCAM; B-CAM; B-CAM cell surface glycoprotein; B-cell adhesion molecule; CD239; F8/G253 antigen; glycoprotein 95kDa; Gplu; LU; lutheran antigen; Lutheran blood group; Lutheran blood group (Auberger b antigen included); Lutheran blood group glycoprotein; Lutheran blood group variant LUGA; lutheran glycoprotein; MSK19 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4059 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P50895 | |
BCAM | |
Recombinant Human BCAM protein (Met1-Ala547). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction