Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547144
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547144 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547144 Supplier Invitrogen™ Supplier No. PA547144
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Approximately 2% cross-reactivity with recombinant human (rh) ALCAM is observed and less than 1% cross-reactivity with rhPECAM, rhEpCAM, rhICAM-1, rhICAM-2, rhICAM-3, and rhVCAM-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAM
Applications Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene BCAM
Gene Accession No. P50895
Gene Alias 1200005K12Rik; antigen identified by monoclonal antibody F8; AU; Auberger B antigen; basal cell adhesion molecule; basal cell adhesion molecule (Lu and Au blood groups); basal cell adhesion molecule (Lutheran blood group); basal cell adhesion molecule Lutheran blood group; BCAM; B-CAM; B-CAM cell surface glycoprotein; B-cell adhesion molecule; CD239; F8/G253 antigen; glycoprotein 95kDa; Gplu; LU; lutheran antigen; Lutheran blood group; Lutheran blood group (Auberger b antigen included); Lutheran blood group glycoprotein; Lutheran blood group variant LUGA; lutheran glycoprotein; MSK19
Gene Symbols BCAM
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human BCAM Glu32-Ala547.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4059
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.