Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCAR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578858
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HEPG2 whole cell. IHC: mouse lymphaden tissue, rat spleen tissue, human tonsil tissue.
Breast tumors are initially dependent on estrogens for growth and progression and can be inhibited by anti-estrogens such as tamoxifen. However, breast cancers progress to become anti-estrogen resistant. Breast cancer anti-estrogen resistance gene 3 was identified in the search for genes involved in the development of estrogen resistance. The gene encodes a component of intracellular signal transduction that causes estrogen-independent proliferation in human breast cancer cells. The protein contains a putative src homology 2 (SH2) domain, a hall mark of cellular tyrosine kinase signaling molecules, and is partly homologous to the cell division cycle protein CDC48.
Specifications
BCAR3 | |
Polyclonal | |
Unconjugated | |
BCAR3 | |
AI131758; And34; AND-34; Bcar3; breast cancer anti-estrogen resistance 3; breast cancer antiestrogen resistance 3 protein; breast cancer anti-estrogen resistance protein 3; dJ1033H22.2 (breast cancer anti-estrogen resistance 3); KIAA0554; novel SH2-containing protein 2; NSP2; OTTHUMP00000011961; p130Cas-binding protein AND-34; SH2 domain-containing protein 3B; SH2D3B; UNQ271/PRO308 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29815, 310838, 8412 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
O75815, Q9QZK2 | |
BCAR3 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human BCAR3 (791-825aa KGAQVNQTERYEKFNQILTALSRKLEPPPVKQAEL). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction