Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104511 Supplier Invitrogen™ Supplier No. PA5104511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BCAR3.

Breast tumors are initially dependent on estrogens for growth and progression and can be inhibited by anti-estrogens such as tamoxifen. However, breast cancers progress to become anti-estrogen resistant. Breast cancer anti-estrogen resistance gene 3 was identified in the search for genes involved in the development of estrogen resistance. The gene encodes a component of intracellular signal transduction that causes estrogen-independent proliferation in human breast cancer cells. The protein contains a putative src homology 2 (SH2) domain, a hall mark of cellular tyrosine kinase signaling molecules, and is partly homologous to the cell division cycle protein CDC48.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAR3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BCAR3
Gene Accession No. O75815
Gene Alias AI131758; And34; AND-34; Bcar3; breast cancer anti-estrogen resistance 3; breast cancer antiestrogen resistance 3 protein; breast cancer anti-estrogen resistance protein 3; dJ1033H22.2 (breast cancer anti-estrogen resistance 3); KIAA0554; novel SH2-containing protein 2; NSP2; OTTHUMP00000011961; p130Cas-binding protein AND-34; SH2 domain-containing protein 3B; SH2D3B; UNQ271/PRO308
Gene Symbols BCAR3
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCAR3(Accession O75815), corresponding to amino acid residues M774-E824.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310838, 8412
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.