Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735068 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5068 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5068 Supplier Proteintech Group Inc Supplier No. 257761AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCAS1 was identified through positional cloning and was found to be overexpressed in most but not all breast cancer cells lines. Little is known about this protein; BCAS1 is cytoplasmically localized and is thought to form homodimers through coiled-coil structures. High levels of BCAS1 mRNA are seen in brain and prostate tissues, with lower amounts observed in colon, intestine and testis. Cells engineered to overexpress BCAS1 did not lose anchorage-dependent growth or increase their rate of growth, suggesting that BCAS1 is not a prototypical oncogene. The BCAS1 gene was also found to be amplified in other carcinomas such as pancreatic carcinoma, suggesting that BCAS1 may play in important role in the control of cell proliferation.

Specifications

Antigen BCAS1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BCAS1
Gene Accession No. O75363, Q80YN3
Gene Alias AIBC1, BCAS1, NABC1
Gene Symbols BCAS1
Host Species Rabbit
Immunogen BCAS1 Fusion Protein Ag22760
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76960, 8537
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.