Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAS2 Antibody, Novus Biologicals™
SDP

Catalog No. NB11040681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11040681 Supplier Novus Biologicals Supplier No. NB11040681
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

BCAS2 Polyclonal specifically detects BCAS2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen BCAS2
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. O75934
Gene Alias breast carcinoma amplified sequence 2, Breast carcinoma-amplified sequence 2, DAM1DNA amplified in mammary carcinoma 1 protein, pre-mRNA-splicing factor SPF27, Snt309, SPF27, spliceosome associated protein, amplified in breast cancer, Spliceosome-associated protein SPF 27
Gene Symbols BCAS2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 175 and the C-terminus (residue 225) of human Breast Cancer Amplified Sequence 2 using the numbering given in entry NP_005863.1 (GeneID 10286).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10286
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.