Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530373 Supplier Invitrogen™ Supplier No. PA530373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat-2, NT2D1, IMR32, MCF-7. Predicted reactivity: Mouse (100%), Rat (98%), Zebrafish (82%), Xenopus laevis (87%), Pig (97%), Chicken (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1.5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene BCAS2
Gene Accession No. O75934, Q9D287
Gene Alias 6430539P16Rik; AI132645; BCAS2; breast carcinoma amplified sequence 2; breast carcinoma-amplified sequence 2; Breast carcinoma-amplified sequence 2 homolog; C76366; C80030; DAM1; DNA amplified in mammary carcinoma 1 protein; pre-mRNA-splicing factor SPF27; Snt309; SPF27; spliceosome associated protein, amplified in breast cancer; spliceosome-associated protein SPF 27
Gene Symbols BCAS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BCAS2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10286, 295334, 68183
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.