Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAS2 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | BCAS2 |
---|---|
Concentration | 0.34 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6289
![]() ![]() |
Proteintech Group Inc
104141AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1. 5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.Specifications
BCAS2 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Non-human Primate, Rat | |
O75934, Q9D287 | |
10286, 295334, 68183 | |
BCAS2 Fusion Protein Ag0669 | |
Primary | |
-20°C | |
BCAS2 |
0.34 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BCAS2, DAM1, Pre mRNA splicing factor SPF27 | |
BCAS2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title