Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596622
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596622 Supplier Invitrogen™ Supplier No. PA596622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAGLVPPGSP GPITRHGSYD SLASDHSGQE DEEWLSQVEI VTHTGPHRRL WMGPQFQFKT IHPSGQTTVI SSSSSVLQSH GPSDTPQPLL DFDTDDLDLN SLRIQPVRSD PVSMPGSSRP VSDRRGVSTV IDAASGTFDR SVTLLEVCGS WPEGFGLRHM SSMEHTEEGL RERLADAMAE SPSRDVVGSG TELQREGSIE TLSNSSGSTS GSIPRNFDGY RSPLPTNESQ PLSLFPTGFP; Positive Samples: HeLa, BT474, Rat brain; Cellular Location: Cytoplasm, Nucleus, cytoskeleton.

BCAS3 is physically associated with histone H3 and histone acetyltransferase complex protein P/CAF (p300/CBP-associated factor) and possesses histone acetyltransferase activity. In breast cancer, several chromosomal sites frequently undergo amplification, implicating the location of genes important for tumor development and progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAS3
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCAS3
Gene Accession No. Q8CCN5, Q9H6U6
Gene Alias 1500019F07Rik; 2610028P08Rik; AU021018; BC028339; Bcas3; BCAS3, microtubule associated cell migration factor; BCAS4/BCAS3 fusion; breast carcinoma amplified sequence 3; breast carcinoma amplified sequence 4/3 fusion protein; breast carcinoma-amplified sequence 3; Breast carcinoma-amplified sequence 3 homolog; GAOB1; K20D4; MAAB; Maab1 protein; metastasis associated antigen of breast cancer; protein Maab1; protein rudhira; RGD1560788; Rudhira
Gene Symbols BCAS3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 674-913 of human BCAS3 (NP_0601493).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192197, 363662, 54828
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.