Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCAS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103669
Description
Antibody detects endogenous levels of total BCAS3.
BCAS3 is physically associated with histone H3 and histone acetyltransferase complex protein P/CAF (p300/CBP-associated factor) and possesses histone acetyltransferase activity. In breast cancer, several chromosomal sites frequently undergo amplification, implicating the location of genes important for tumor development and progression.
Specifications
BCAS3 | |
Polyclonal | |
Unconjugated | |
BCAS3 | |
1500019F07Rik; 2610028P08Rik; AU021018; BC028339; Bcas3; BCAS3, microtubule associated cell migration factor; BCAS4/BCAS3 fusion; breast carcinoma amplified sequence 3; breast carcinoma amplified sequence 4/3 fusion protein; breast carcinoma-amplified sequence 3; Breast carcinoma-amplified sequence 3 homolog; GAOB1; K20D4; MAAB; Maab1 protein; metastasis associated antigen of breast cancer; protein Maab1; protein rudhira; RGD1560788; Rudhira | |
Rabbit | |
Affinity chromatography | |
RUO | |
192197, 54828 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8CCN5, Q9H6U6 | |
BCAS3 | |
A synthesized peptide derived from human BCAS3(Accession Q9H6U6), corresponding to amino acid residues L692-P742. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction