Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAS3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8782 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8782 Supplier Bethyl Laboratories, Inc Supplier No. A300916A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 or Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse

The Breast carcinoma amplified sequence (BCAS3) gene is localized to 17q23, a region that is often amplified in breast cancers. Copy number gains at this region have also been reported in other tumors such as brain, lung, liver, testis and bladder. BCAS3 is thought to be an estrogen receptor (ER)-alpha co-activator that acts through PELP1, another ER-alpha that in turn activates BCAS3 expression. This may set up a positive feedback loop leading to ER-a signal amplification in the cell which may play a significant role in breast cancer. BCAS3 expression has also been observed in embryonic stem cells and vascular precursors, suggesting that it may also play a role in embryogenesis and tumor angiogenesis.

Specifications

Antigen BCAS3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene BCAS3
Gene Accession No. Q9H6U6
Gene Alias Breast carcinoma-amplified sequence 3; GAOB1; Maab1 protein; BCAS3
Gene Symbols BCAS3
Host Species Rabbit
Immunogen Between 870 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54828
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.