Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558549 Supplier Invitrogen™ Supplier No. PA558549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVALLMLLVD ADQPEPMRSG ARELALFLTP EPGAEAKEVE ETIEGMLLRL EEFCSLADLI RSDTSQILEE NIPVLKAKLT EMRGIYAKVD RL Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 29%.

The Breast carcinoma amplified sequence (BCAS4) gene is localized to 20q13, a region that is often amplified in breast cancers. It was isolated by the cloning of a highly overexpressed 1.3 kb transcript from the fusion of the last two exons of the BCAS3 and BCAS4 genes. This BCAS3-BCAS4 fusion transcript is only found in the MCF7 cell line, but the BCAS4 gene is also overexpressed in nine of thirteen breast cancer cell lines. The role of BCAS4 is unknown but its mRNA is highly expressed in thymus, spleen, kidney, brain, and placenta, suggesting BCAS4 plays other roles in multiple tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAS4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCAS4
Gene Accession No. Q8TDM0
Gene Alias BCAS4; BCAS4/BCAS3 fusion; breast carcinoma amplified sequence 4; breast carcinoma amplified sequence 4/3 fusion protein; breast carcinoma-amplified sequence 4; CNOL; RP5-1071L10.2
Gene Symbols BCAS4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCAS4. Recombinant protein control fragment (Product #RP-95486).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55653
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.