Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAT1 Antibody (3C8), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259430G
This item is not returnable.
View return policy
Description
BCAT1 Monoclonal antibody specifically detects BCAT1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
BCAT1 | |
Monoclonal | |
DyLight 488 | |
BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39 | |
Recombinant human BCAT1 (1-386aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
586 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
3C8 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction