Learn More
Description
Specifications
Specifications
| Antigen | BCAT1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q8CBC8 |
| Gene Alias | BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39 |
| Gene Symbols | BCAT1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
