Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAT1 Monoclonal Antibody (OTI3F5)
GREENER_CHOICE

Catalog No. PIMA525892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525892 Supplier Invitrogen™ Supplier No. MA525892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCAT1 Monoclonal Antibody for IHC (P), Western Blot

This gene encodes the cytosolic form of the enzyme branched-chain amino acid transaminase. This enzyme catalyzes the reversible transamination of branched-chain alpha-keto acids to branched-chain L-amino acids essential for cell growth. Two different clinical disorders have been attributed to a defect of branched-chain amino acid transamination: hypervalinemia and hyperleucine-isoleucinemia. As there is also a gene encoding a mitochondrial form of this enzyme, mutations in either gene may contribute to these disorders. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3F5
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Bcat1
Gene Accession No. P24288, P54687
Gene Alias BCAT(c); Bcat1; BCATC; BCT1; branched chain amino acid transaminase 1; branched chain amino acid transaminase 1, cytosolic; branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; branched-chain-amino-acid aminotransferase, cytosolic; cytosolic branched-chain amino acid aminotransferase; Eca39; MECA39; placental protein 18; PNAS121; PP18; Protein ECA39
Gene Symbols Bcat1
Host Species Mouse
Immunogen Full length human recombinant protein of BCAT1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12035, 486633, 586
Target Species Canine, Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.