Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578859
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578859 Supplier Invitrogen™ Supplier No. PA578859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Raji whole cell, human MOLT-4 whole cell. IHC: human ovarian adenoma tissue, human pancreatic cancer tissue.

This gene encodes the cytosolic form of the enzyme branched-chain amino acid transaminase. This enzyme catalyzes the reversible transamination of branched-chain alpha-keto acids to branched-chain L-amino acids essential for cell growth. Two different clinical disorders have been attributed to a defect of branched-chain amino acid transamination: hypervalinemia and hyperleucine-isoleucinemia. As there is also a gene encoding a mitochondrial form of this enzyme, mutations in either gene may contribute to these disorders. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Bcat1
Gene Accession No. P54687
Gene Alias BCAT(c); Bcat1; BCATC; BCT1; branched chain amino acid transaminase 1; branched chain amino acid transaminase 1, cytosolic; branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; branched-chain-amino-acid aminotransferase, cytosolic; cytosolic branched-chain amino acid aminotransferase; Eca39; MECA39; placental protein 18; PNAS121; PP18; Protein ECA39
Gene Symbols Bcat1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human BCAT1 (360-375aa KLASRILSKLTDIQYGR).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 586
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.