Learn More
Invitrogen™ BCAT1 Polyclonal Antibody

Description
Positive test controls include: Jurkat, Raji, SH-SY5Y, Neuro-2a, Mouse brain. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: EVVGTFKAKD LIVTPATILK EKPDPNNLVF GTVFTDHMLT VEWSSEFGWE KPHIKPLQNL SLHPGSSALH YAVELFEGLK AFRGVDNKIR LFQPNLNMDR MYRSAVRATL PVFDKEELLE CIQQLVKLDQ EWVPYSTSAS LYIRPTFIGT EPSLGVKKPT KALLFVLLSP V.
Specifications
Specifications
| Antigen | BCAT1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.29 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Bcat1 |
| Gene Accession No. | P24288, P54687, P54690 |
| Gene Alias | BCAT(c); Bcat1; BCATC; BCT1; branched chain amino acid transaminase 1; branched chain amino acid transaminase 1, cytosolic; branched chain amino-acid transaminase 1, cytosolic; branched chain aminotransferase 1, cytosolic; branched-chain-amino-acid aminotransferase, cytosolic; cytosolic branched-chain amino acid aminotransferase; Eca39; MECA39; placental protein 18; PNAS121; PP18; Protein ECA39 |
| Gene Symbols | Bcat1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.