Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAT2 Antibody (7G3A11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | BCAT2 |
---|---|
Clone | 7G3A11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61699
![]() |
Novus Biologicals
NBP261699 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
BCAT2 Monoclonal specifically detects BCAT2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
BCAT2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
BCAM, BCAT(m), BCATM, BCT2EC 2.6.1.42, branched chain amino-acid transaminase 2, mitochondrial, branched chain aminotransferase 2, mitochondrial, branched-chain-amino-acid aminotransferase, mitochondrial, ECA40, Placental protein 18, PP18 | |
BCAT2 | |
IgG1 | |
Protein G purified |
7G3A11 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
metabolism | |
587 | |
Purified recombinant fragment of human BCAT2 (AA: 259-393) expressed in E. Coli. | |
Primary | |
44.3 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title