Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAT2 Mouse anti-Human, Clone:7G3A11, Abnova™

Catalog No. 89333909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-909 Supplier Abnova Corporation Supplier No. MAB17522
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human BCAT2.

This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine, isoleucine, and valine. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen BCAT2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7G3A11
Conjugate Unconjugated
Description branched chain aminotransferase 2, mitochondrial
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene BCAT2
Gene Alias BCAM, BCT2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 259-393 of human BCAT2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 587
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.