Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562914 Supplier Invitrogen™ Supplier No. PA562914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAALGQIW ARKLLSVPWL LCGPRRYASS SFKAADLQLE MTQKPHKKPG PGEPLVFGKT FTDHML Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.

This gene encodes a branched chain aminotransferase found in mitochondria. The encoded protein forms a dimer that catalyzes the first step in the production of the branched chain amino acids leucine, isoleucine, and valine. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCAT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCAT2
Gene Accession No. O15382
Gene Alias BCAM; BCAT(m); Bcat2; Bcat-2; Bcatm; BCT2; branched chain amino acid transaminase 2; branched chain amino acid transaminase 2, mitochondrial; branched chain amino-acid transaminase 2, mitochondrial; branched chain aminotransferase 2, mitochondrial; branched-chain-amino-acid aminotransferase, mitochondrial; Eca40; heart branched chain aminotransferase precursor encoding mitochondrial protein; mBcat; placental protein 18; PP18
Gene Symbols BCAT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCAT2. Recombinant protein control fragment (Product #RP-98983).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.