Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCCIP Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122994FR
Description
BCCIP Polyclonal antibody specifically detects BCCIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BCCIP | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BCCIPalpha, BRCA2 and CDKN1A interacting protein, BRCA2 and CDKN1A-interacting protein, BRCA2 and Cip1/p21 interacting protein, cdk inhibitor p21 binding protein, P21- and CDK-associated protein 1, Protein TOK-1, TOK-1, TOK-1alpha, TOK1BCCIPbeta, TOK-1beta | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human BRCA2 and CDKN1A interacting protein using the numbering given in entry NP_510868.1 (GeneID 56647). | |
0.1 mL | |
Cell Cycle and Replication | |
56647 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction