Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCDIN3D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116400
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116400 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116400 Supplier Invitrogen™ Supplier No. PA5116400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCDIN3D Polyclonal Antibody for Western Blot

O-methyltransferase that specifically monomethylates 5'- monophosphate of cytoplasmic histidyl tRNA (tRNA(His)), acting as a capping enzyme by protecting tRNA(His) from cleavage by DICER1. Also able, with less efficiently, to methylate the 5' monophosphate of a subset of pre- miRNAs, acting as a negative regulator of miRNA processing. The 5' monophosphate of pre-miRNAs is recognized by DICER1 and is required for pre-miRNAs processing: methylation at this position reduces the processing of pre-miRNAs by DICER1. Was also reported to mediate dimethylation of pre-miR-145; however dimethylation cannot be reproduced by another group which observes a monomethylation of pre-miR-145.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCDIN3D
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BCDIN3D
Gene Accession No. Q7Z5W3
Gene Alias BCDIN3 domain containing RNA methyltransfease; BCDIN3 domain containing RNA methyltransferase; BCDIN3 domain-containing protein; BCDIN3D; pre-miRNA 5'-monophosphate methyltransferase; probable methyltransferase BCDIN3D; RNA 5'-monophosphate methyltransferase
Gene Symbols BCDIN3D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 243-276 amino acids from human BCDIN3D.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.