Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCDIN3D Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304633100UL
Description
BCDIN3D Polyclonal specifically detects BCDIN3D in Human, Mouse samples. It is validated for Western Blot.Specifications
BCDIN3D | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
144233 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
BCDIN3 domain containing, BCDIN3 domain-containing protein, EC 2.1.1, EC 2.1.1.-, probable methyltransferase BCDIN3D | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-292 of human BCDIN3D (NP_859059.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction