Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCHE Mouse anti-Human, Clone: 1E8, Bioss
SDP

Catalog No. 501979894
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9894 Supplier Bioss Supplier No. BSM2011M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.

Specifications

Antigen BCHE
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E8
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BCHE
Gene Accession No. P06276
Gene Alias Acylcholine acylhydrolase; BCHE; BtChE; butylcholinesterase; Butyrylcholine esterase; butyrylcholinesterase; C730038G20Rik; CHE1; CHE2; Choline esterase II; Cholinesterase; cholinesterase (serum) 2; cholinesterase 1; E1; EC 3.1.1.8; Pseudocholinesterase
Gene Symbols BCHE
Host Species Mouse
Immunogen Full length native protein purified from human.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 590
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.