Learn More
BCHE Mouse anti-Human, Clone: OTI4C12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502288

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mutant alleles at the BCHE locus are responsible for suxamethonium sensitivity. Homozygous persons sustain prolonged apnea after administration of the muscle relaxant suxamethonium in connection with surgical anesthesia. The activity of pseudocholinesterase in the serum is low and its substrate behavior is atypical. In the absence of the relaxant, the homozygote is at no known disadvantage.Specifications
BCHE | |
Monoclonal | |
Unconjugated | |
BCHE | |
CHE1, CHE2, E1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
590 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4C12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06276 | |
BCHE | |
Full length human recombit protein of human BCHE produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.