Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCKDH Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206192
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6192 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6192 Supplier Fabgennix Inc Supplier No. BCKD101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCKDH Polyclonal antibody specifically detects BCKDH in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen BCKDH
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene BCKDK
Gene Accession No. O14874, O55028, Q00972
Gene Alias [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial; AI327402; BCKDHKIN; BCKDK; BCKDKD; BCKD-kinase; BDK; branched chain alpha-ketoacid dehydrogenase kinase; branched chain ketoacid dehydrogenase kinase; branched-chain alpha-ketoacid dehydrogenase kinase; EC 2.7.11.4
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 170-200 on human BCKD protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10295, 12041, 29603
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.