Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCKDHA Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297503AF594
Description
BCKDHA Polyclonal specifically detects BCKDHA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
BCKDHA | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial, BCKDE1A, BCKDH E1-alpha, branched chain keto acid dehydrogenase E1, alpha polypeptide, branched chain keto acid dehydrogenase E1, alpha polypeptide (maple syrup urinedisease), Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain, EC 1.2.4.4,2-oxoisovalerate dehydrogenase (lipoamide), FLJ45695, MSU, MSUD1, OVD1A | |
Produced in rabbits immunized with E. coli-derived Human BCKDHA fragment. | |
0.1 mL | |
Phospho Specific | |
593 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction