Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCKDHA Antibody (BLR060F), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314681
This item is not returnable.
View return policy
Description
BCKDHA Monoclonal antibody specifically detects BCKDHA in Human, Mouse samples. It is validated for Western Blot, Western Blot, ImmunoprecipitationSpecifications
BCKDHA | |
Monoclonal | |
Unconjugated | |
2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial, BCKDE1A, BCKDH E1-alpha, branched chain keto acid dehydrogenase E1, alpha polypeptide, branched chain keto acid dehydrogenase E1, alpha polypeptide (maple syrup urinedisease), Branched-chain alpha-keto acid dehydrogenase E1 component alpha chain, EC 1.2.4.4,2-oxoisovalerate dehydrogenase (lipoamide), FLJ45695, MSU, MSUD1, OVD1A | |
surrounding serine 292 | |
RUO | |
593 | |
Store at 2 - 8°C/ 1 year from date of receipt |
Western Blot, Immunoprecipitation | |
BLR060F | |
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA and 0.09% Sodium Azide | |
Rabbit | |
100 μg | |
Primary | |
Human, Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction