Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCKDHA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597248 Supplier Invitrogen™ Supplier No. PA597248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SEQYRGDGIA ARGPGYGIMS IRVDGNDVFA VYNATKEARR RAVAENQPFL IEAMTYRIGH HSTSDDSSAY RSVDEVNYWD KQDHPISRLR HYLLSQGWWD EEQEKAWRKQ SRRKVMEAFE QAERKPKPNP NLLFSDVYQE MPAQLRKQQE SLARHLQTYG EHYPLDHFDK; Positive Samples: NCI-H460, LO2, DU145, NIH/3T3, Mouse liver, Mouse kidney, Mouse heart, Rat kidney, Rat liver; Cellular Location: Mitochondrion matrix.

The BCKDHA gene encodes the E1-alpha subunit of the branched-chain alpha-keto acid (BCAA) dehydrogenase complex (BCKD; EC 1.2.4.4), an inner-mitochondrial enzyme complex that catalyzes the oxidative decarboxylation of the branched-chain alpha-ketoacids derived from isoleucine, leucine, and valine. This reaction is the second major step in the catabolism of the branched-chain amino acids (Wynn et al., 1998). The BCKD complex consists of 3 catalytic components: a heterotetrameric (alpha2-beta2) branched-chain alpha-keto acid decarboxylase (E1), a homo-24-meric dihydrolipoyl transacylase (E2), and a homodimeric dihydrolipoamide dehydrogenase (E3). E1 is a thiamine pyrophosphate (TPP)-dependent enzyme. The reaction is irreversible and constitutes the first committed step in BCAA oxidation. The BCKDHB gene encodes the beta subunit of E1. The complex also contains 2 regulatory enzymes, a kinase and a phosphorylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCKDHA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCKDHA
Gene Accession No. P11960, P12694, P50136
Gene Alias 2-oxoisovalerate dehydrogenase (lipoamide); 2-oxoisovalerate dehydrogenase subunit alpha, mitochondrial; BCKAD E1[a]; BCKDA; BCKDE1A; BCKDH E1-alpha; BCKDHA; branched chain alpha-ketoacid dehydrogenase subunit E1 alpha; branched chain keto acid dehydrogenase E1 alpha protein; branched chain keto acid dehydrogenase E1, alpha polypeptide; branched chain keto acid dehydrogenase subunit E1, alpha polypeptide; branched chain ketoacid dehydrogenase E1, alpha polypeptide; branched-chain alpha-keto acid dehydrogenase E1 component alpha chain; E1a; MSU; MSUD1; OVD1A
Gene Symbols BCKDHA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 276-445 of human BCKDHA (NP_0007001).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12039, 25244, 593
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.