Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCKDHB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587963 Supplier Thermo Scientific Supplier No. PA587963
Add to Cart
Add to Cart

BCKDHB Polyclonal Antibody

BCKDHB Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen BCKDHB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Bckdhb
Gene Accession No. P35738, Q6P3A8
Gene Alias 2-oxoisovalerate dehydrogenase beta subunit; 2-oxoisovalerate dehydrogenase subunit beta, mitochondrial; BCKD BCKDH E1-beta; BCKDE1B; BCKDH E1-beta; BCKDHB; branched chain alpha-ketoacid dehydrogenase E1-beta subunit; branched chain keto acid dehydrogenase E1 beta; branched chain keto acid dehydrogenase E1 subunit beta; branched chain keto acid dehydrogenase E1, beta polypeptide; branched chain ketoacid dehydrogenase E1, beta polypeptide; branched-chain alpha-keto acid dehydrogenase E1 component beta chain; dJ279A18.1; E1B; E1b-beta subunit of the branched-chain complex; testis secretory sperm-binding protein Li 240mP
Gene Symbols Bckdhb
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 183-392 of human BCKDHB (NP_8988711).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12040, 29711
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.