Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCKDK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553697 Supplier Invitrogen™ Supplier No. PA553697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLDDHKDVVT LLAEGLRESR KHIEDEKLVR YFLDKTLTSR LGIRMLATHH LALHEDKPDF VGIICTRLSP KKIIEKWVDF ARRLCEHKYG NAPRVRINGH VAARFPFIPM PLDY Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.

The second major step in the catabolism of the branched-chain amino acids, isoleucine, leucine, and valine, is irreversibly catalyzed by the branched-chain alpha-keto acid dehydrogenase complex (BCKD), an inner-mitochondrial enzyme complex composed of 3 catalytic components: a branched-chain alpha-keto acid decarboxylase (E1), a dihydrolipoyl transacylase (E2), and a dihydrolipoamide dehydrogenase (E3). The complex also contains 2 enzymes that regulated the state of activity of the BCKD complex: a kinase (BCKDK), and a phosphorylase. The ubiquitously expressed kinase contains 1 histidine kinase domain. Maple syrup urine disease (MSUD) is a pathology secondary to an enzyme defect in the catabolic pathway of leucine, isoleucine, and valine. Accumulation of these amino acids and their corresponding keto acids results in encephalopathy and progressive neurodegeneration in infants not treated for MSUD.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCKDK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCKDK
Gene Accession No. O14874, O55028, Q00972
Gene Alias [3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial; AI327402; BCKDHKIN; BCKDK; BCKDKD; BCKD-kinase; BDK; branched chain alpha-ketoacid dehydrogenase kinase; branched chain ketoacid dehydrogenase kinase; branched-chain alpha-ketoacid dehydrogenase kinase; EC 2.7.11.4
Gene Symbols BCKDK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCKDK. Recombinant protein control fragment (Product #RP-91406).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10295, 12041, 29603
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.