Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-10 Antibody (OTI4A8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271398F
Description
Bcl-10 Monoclonal specifically detects Bcl-10 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Bcl-10 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
BCL10 | |
Full length recombinant protein of human Bcl-10 (NP_003912) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4A8 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer | |
8915.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction