Learn More
Bcl-10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA585359
Description
Keep as concentrated solution. Predicted reactivity: Mouse (95%), Rat (96%), Pig (96%), Rhesus Monkey (99%), Bovine (99%). Positive Control: Jurkat, Neuro2A, GL261, C8D30. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
Specifications
Bcl-10 | |
Polyclonal | |
Unconjugated | |
BCL10 | |
AI132454, B cell CLL/lymphoma 10, B cell leukemia/lymphoma 10, B-cell CLL/lymphoma 10, B-cell leukemia/lymphoma 10, B-cell lymphoma/leukemia 10, BCL10, Bcl-10, BCL10 immune signaling adaptor, C81403, CARD containing molecule enhancing NF-kB, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, cE10, c-E10, CED-3/ICH-1 prodomain homologous E10-like regulator, cellular homolog of vCARMEN, Cellular-E10, CIPER, Clap, hCLAP, IMD37, Mamm, mammalian CARD-containing adapter molecule E10, mCIPER, mCLAP, mE10, OTTHUMP00000011647, OTTHUMP00000036080, RCD, RP11-234D19.2, R-RCD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12042, 83477, 8915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal, pH 7 | |
O95999, Q9QYN5, Q9Z0H7 | |
BCL10 | |
Recombinant protein encompassing a sequence within the center region of human BCL10. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.