Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-2 Antibody (8C8), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233313MFV450
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human, Porcine, Monkey, Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Bcl-2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Pig, Monkey, Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
8C8 | |
Flow Cytometry | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
A synthetic peptide, aa41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein. (Uniprot: P10415 ) | |
0.1 mL | |
Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Mitophagy, Oncogenes, Phospho Specific, Tumor Suppressors | |
596 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction