Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP230108SS
Description
Bcl-2 Polyclonal specifically detects Bcl-2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Bcl-2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
Rabbit | |
26 kDa | |
0.025 mg | |
Primary | |
Human Bcl-2 has 2 isoforms, including isoform 1 (239aa, 26kD), and isoform 2 (205aa, 22kD). Mouse Bcl-2 has 2 isoforms, including isoform 1 (236aa, 26kD), and isoform 2 (199aa, 22kD). Rat Bcl-2 has one isoform (236aa, 27kD). NBP2-30108 can detect all isoforms of human, mouse and rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2ug/ml, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 10ug/ml, Immunohistochemistry-Paraffin 2ug/ml | |
AAH27258 | |
BCL2 | |
Antibody was raised against a peptide corresponding to 15 amino acids near the N terminus of human Bcl-2. The immunogen is located within the first 50 amino acids of Bcl-2. Amino Acid Squence: KYIHYKLSQRGYEWD | |
Ion exchange chromatography | |
Apoptosis, Autophagy, Cancer, Cellular Markers, Phospho Specific, Tumor Suppressors | |
596 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction