Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-2 Antibody (SPM530), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234442DL594
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Bcl-2 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BCL2 | |
A synthetic peptide, aa41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein (Uniprot: P10415) | |
0.1 mL | |
Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Mitophagy, Oncogenes, Phospho Specific, Tumor Suppressors | |
This antibody recognizes a protein of 25-26kDa, identified as the Bcl-2 lpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of Bcl-2 lpha oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2 lpha protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express Bcl-2 protein and the small number in which the neoplastic cells are Bcl-2 negative. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM530 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction